Learn More
Cole-Parmer Cylindrical Dewar Flask with Handle, 1000 mL
Supplier: Cole-Parmer 0377115
These Dewars feature an exterior metal housing with a boroscilicate glass interior. There are no pads between walls to eliminate heat transfer potential. These are ideal for temporary storage or for transfer applications. Dewar flasks come with lids.