Learn More
Synthware™ Dewer Style Vacuum Trap
GREENER_CHOICESDP
Catalog No. 31501847 Shop All Kemtech Products
Change view
Click to view available options
Type:
Glass Stopcock
PTFE Stopcock
Vacuum Trap - Dewar
Capacity (Metric):
250 mL
500 mL
1000 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Capacity (Metric)
31-501-847 PTFE Stopcock 1000 mL
31-501-842 Glass Stopcock 250 mL
31-501-843 PTFE Stopcock 250 mL
31-501-844 Vacuum Trap - Dewar 500 mL
31-501-845 PTFE Stopcock 500 mL
31-501-846 Glass Stopcock 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 31-501-847 Supplier Synthware™ Supplier No. V211000
Add to Cart
Add to Cart
Dry ice, dewar syle trap condenser designed for large volume solvent removal
Specifications
For Use With (Application) Organic Synthesis
Type PTFE Stopcock
Material Borosilicate Glass
Capacity (Metric) 1000 mL
Quantity 1 Ea.