Learn More
Synthware™ Dewer Style Vacuum Trap- Microscale
GREENER_CHOICESDP
Catalog No. 31501841 Shop All Kemtech Products
Change view
Click to view available options
Type:
Vacuum Trap - Dewar
Catalog No. Type
31-501-841 Vacuum Trap - Dewar
1 options
Catalog No. 31-501-841 Supplier Synthware™ Supplier No. V200100
Add to Cart
Add to Cart
Mircoscale dry ice, dewar style trap
Specifications
For Use With (Application) Organic Synthesis
Type Vacuum Trap - Dewar
Material Borosilicate Glass
Capacity (Metric) 100 mL
Quantity 1 Ea.