Learn More
Synthware™ Dewer Style Vacuum Trap
GREENER_CHOICESDP
Catalog No. 31501842 Shop All Kemtech Products
Change view
Click to view available options
Capacity (Metric):
250 mL
Catalog No. Capacity (Metric)
31-501-842 250 mL
1 options
Catalog No. 31-501-842 Supplier Synthware™ Supplier No. V210250G
Add to Cart
Add to Cart
Dry ice, dewar syle trap condenser designed for large volume solvent removal
Specifications
For Use With (Application) Organic Synthesis
Type Vacuum Trap - Dewar
Material Borosilicate Glass
Capacity (Metric) 250 mL
Quantity 1 Ea.