Learn More
MilliporeSigma™ Supelco™ DEX™ GC column Screening Kit I
Catalog No. 111006054 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Application):
Gas Chromatography
Catalog No. For Use With (Application)
11-100-6054 Gas Chromatography
1 options
Catalog No. 11-100-6054 Supplier MilliporeSigma™ Supelco™ Supplier No. 24340
Add to Cart
Add to Cart
Kit provides the tools you need to perform most chiral separations
Confirm identities of enantiomers by monitoring elution order changes (enantioreversal) from one column to another. In combination, the columns in the two kits span the full range of Supelco DEX column enantioselectivity.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Gas Chromatography
Includes One 30 m x 0.25 mm I.D., 0.25 μm column of each type: α-DEX 120, β-DEX 120 and γ-DEX 120
DEX is a trademark of Sigma-Aldrich Co. LLC