Learn More
Medchemexpress LLC DEX-maleimide | 2754462-35-4 | 99.8% | C29H36FN3O7 | 1 ML
SDP
Catalog No. 5000357873
Encompass_Preferred
DEX-maleimide is a coupling agent designed to synthesize conjugates containing single-domain antibodies (VHH) conjugated with antigens and anti-inflammatory agents. It finds application in research related to autoimmune diseases.
  • Acts as a coupling agent.
  • Synthesizes conjugates with single-domain antibodies (VHH).
  • Facilitates conjugation of antigens.
  • Supports conjugation of anti-inflammatory agents.
  • Useful for research on autoimmune diseases.
Catalog No. 50-003-57873 Supplier Medchemexpress LLC Supplier No. HY4622510MM/1ML
DEX-maleimide is a coupling agent designed to synthesize conjugates containing single-domain antibodies (VHH) conjugated with antigens and anti-inflammatory agents. It finds application in research related to autoimmune diseases.
  • Acts as a coupling agent.
  • Synthesizes conjugates with single-domain antibodies (VHH).
  • Facilitates conjugation of antigens.
  • Supports conjugation of anti-inflammatory agents.
  • Useful for research on autoimmune diseases.