Learn More
Medchemexpress LLC DEX-maleimide | 2754462-35-4 | 99.8% | C29H36FN3O7 | 25 MG
SDP
Catalog No. 5000358960
Encompass_Preferred
DEX-maleimide is a coupling agent used to synthesize conjugates containing single-domain antibodies (VHH) conjugated with antigens and anti-inflammatory agents. It can be used for research on autoimmune diseases.
  • Coupling agent
  • Synthesizes conjugates containing single-domain antibodies (VHH)
  • Synthesizes conjugates with antigens and anti-inflammatory agents
  • Applicable for research on autoimmune diseases
Catalog No. 50-003-58960 Supplier Medchemexpress LLC Supplier No. 25MGHY46225
DEX-maleimide is a coupling agent used to synthesize conjugates containing single-domain antibodies (VHH) conjugated with antigens and anti-inflammatory agents. It can be used for research on autoimmune diseases.
  • Coupling agent
  • Synthesizes conjugates containing single-domain antibodies (VHH)
  • Synthesizes conjugates with antigens and anti-inflammatory agents
  • Applicable for research on autoimmune diseases