Learn More
Medchemexpress LLC DEX-maleimide | 2754462-35-4 | 99.8% | C29H36FN3O7 | 50 MG
SDP
Catalog No. 5000358221
Encompass_Preferred
DEX-maleimide is a coupling agent that can be used to synthesize conjugates containing single-domain antibodies (VHH) conjugated with antigens and anti-inflammatory agents. It is also applicable for research concerning autoimmune diseases.
  • Used to synthesize conjugates containing single-domain antibodies (VHH) with antigens and anti-inflammatory agents.
  • Applicable for research concerning autoimmune diseases.
Catalog No. 50-003-58221 Supplier Medchemexpress LLC Supplier No. HY4622550MG
DEX-maleimide is a coupling agent that can be used to synthesize conjugates containing single-domain antibodies (VHH) conjugated with antigens and anti-inflammatory agents. It is also applicable for research concerning autoimmune diseases.
  • Used to synthesize conjugates containing single-domain antibodies (VHH) with antigens and anti-inflammatory agents.
  • Applicable for research concerning autoimmune diseases.