Learn More
YSI 3P LifeScience 2356 Glucose Standard 500 mg/dL (5 g/L)
Catalog No. 50200152
Encompass
2356 Glucose Standard 500 mg/dL (5 g/L). 500mg/dL glucose standard solution for use with YSI 2300, 2700, 2900 Series and 7100 analyzers (125mL). Minimum order quantity 2.
Catalog No. 50-200-152 Supplier YSI 3P LifeScience Supplier No. 2356
2356 Glucose Standard 500 mg/dL (5 g/L). 500mg/dL glucose standard solution for use with YSI 2300, 2700, 2900 Series and 7100 analyzers (125mL). Minimum order quantity 2.