Learn More
Cole-Parmer Idex U-467BLK Port Plug, Short Head/Standard Knurl, Black Acetal, 10-32 Coned; 1/EA
Catalog No. NC1766040 Shop All Cole-Parmer Products
Encompass
Use the plugs to close off unused ports in valves and multiport connectors. Select coned plugs for high-pressure applications; flat-bottom plugs for low-pressure applications. Color-coded plugs are ideal for easily identifying stored columns that have different packing materials.
Catalog No. NC1766040 Supplier Cole-Parmer Supplier No. 0201853
Use the plugs to close off unused ports in valves and multiport connectors. Select coned plugs for high-pressure applications; flat-bottom plugs for low-pressure applications. Color-coded plugs are ideal for easily identifying stored columns that have different packing materials.