Learn More
Apexbio Technology LLC Dexamethasone Sodium Phosphate 55203-24-2 1g
SDP
Catalog No. 5000063717
Encompass_Preferred
Dexamethasone Sodium Phosphate is a synthetic glucocorticoid compound characterized by potent anti-inflammatory and immunomodulatory properties Mechanistically it functions primarily through the modulation of interleukin receptor signaling pathways and suppression of cyclooxygenase-2 (COX-2) activity By attenuating cytokine-mediated responses at these molecular targets dexamethasone sodium phosphate serves as an inhibitor in immune cell signaling and inflammatory cascades In biomedical research settings this agent is frequently utilized in studies investigating immune system regulation inflammatory diseases cytokine expressions and glucocorticoid receptor-mediated gene regulation pathways Additionally it is employed experimentally to elucidate mechanisms underlying glucocorticoid responsiveness and resistance at cellular and molecular levels
Catalog No. 50-000-63717 Supplier Apexbio Technology LLC Supplier No. B15881000
Dexamethasone Sodium Phosphate is a synthetic glucocorticoid compound characterized by potent anti-inflammatory and immunomodulatory properties Mechanistically it functions primarily through the modulation of interleukin receptor signaling pathways and suppression of cyclooxygenase-2 (COX-2) activity By attenuating cytokine-mediated responses at these molecular targets dexamethasone sodium phosphate serves as an inhibitor in immune cell signaling and inflammatory cascades In biomedical research settings this agent is frequently utilized in studies investigating immune system regulation inflammatory diseases cytokine expressions and glucocorticoid receptor-mediated gene regulation pathways Additionally it is employed experimentally to elucidate mechanisms underlying glucocorticoid responsiveness and resistance at cellular and molecular levels