Learn More
Cayman Chemical Dexamethasone 9,11-epoxide, Purity: ≥98%, CAS: 24916-90-3, 1 g
SDP
Catalog No. 502913192
Encompass_Preferred
A potential impurity found in preparations of dexamethasone; an autooxidative degradation product formed under alkaline conditions
Catalog No. 50-291-3192 Supplier Cayman Chemical Supplier No. 361611 G
A potential impurity found in preparations of dexamethasone; an autooxidative degradation product formed under alkaline conditions