Learn More
Cayman Chemical Dexamethasone 9,11-epoxide, Purity: ≥98%, CAS: 24916-90-3, 100 mg
SDP
Catalog No. 502909143
Encompass_Preferred
A potential impurity found in preparations of dexamethasone; an autooxidative degradation product formed under alkaline conditions
Catalog No. 50-290-9143 Supplier Cayman Chemical Supplier No. 36161100 MG
A potential impurity found in preparations of dexamethasone; an autooxidative degradation product formed under alkaline conditions