Learn More
Cayman Chemical Dexamethason-d4 10mg
SDP
Catalog No. 502986198
Encompass_Preferred
An internal standard for the quantification of dexamethasone by GC- or LC-MS
Catalog No. 50-298-6198 Supplier Cayman Chemical Supplier No. 2210010 MG
Add to Cart
Add to Cart
An internal standard for the quantification of dexamethasone by GC- or LC-MS