Learn More
Cayman Chemical Dexamethasone-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 2483831-63-4, 5 mg
SDP
Catalog No. 502987290
Encompass_Preferred
An internal standard for the quantification of dexamethasone by GC- or LC-MS
Catalog No. 50-298-7290 Supplier Cayman Chemical Supplier No. 221005 MG
An internal standard for the quantification of dexamethasone by GC- or LC-MS