Learn More
Cayman Chemical Dexamethasone Palmitate, Purity: ≥98%, CAS: 14899-36-6, 10 mg
SDP
Catalog No. 502913965
Encompass_Preferred
A prodrug form of dexamethasone; binds to the glucocorticoid receptor; reduces LPS-induced increases in cytokine levels in RAW 264.7 macrophages at 100 µg/ml in lipid nanoparticles; reduces paw inflammation and edema in a mouse model of collagen-induced rheumatoid arthritis at 1 mg/kg; inhibits VEGF-induced vascular hyperpermeability for up to nine months in rabbits at 1,280/animal as an intravitreal emulsion injection
Catalog No. 50-291-3965 Supplier Cayman Chemical Supplier No. 3545110 MG
A prodrug form of dexamethasone; binds to the glucocorticoid receptor; reduces LPS-induced increases in cytokine levels in RAW 264.7 macrophages at 100 µg/ml in lipid nanoparticles; reduces paw inflammation and edema in a mouse model of collagen-induced rheumatoid arthritis at 1 mg/kg; inhibits VEGF-induced vascular hyperpermeability for up to nine months in rabbits at 1,280/animal as an intravitreal emulsion injection