Learn More
Sigma Aldrich Fine Chemicals Biosciences Dexamethasone, 97, 50-02-2, MFCD00064136, 100mg
Catalog No. 501657009
Encompass
Empirical Formula (Hill Notation) C22H29FO5, ≥97%, Molecular Weight 392.46, mp 262-264 °C (lit.), powder, BioReagent, Synonym: (11β,16α)-9-Fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione, 9α-Fluoro-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, 9α-Fluoro-16α-methylprednisolone, Prednisolone F
Catalog No. 50-165-7009 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D4902100MG
Empirical Formula (Hill Notation) C22H29FO5, ≥97%, Molecular Weight 392.46, mp 262-264 °C (lit.), powder, BioReagent, Synonym: (11β,16α)-9-Fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione, 9α-Fluoro-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, 9α-Fluoro-16α-methylprednisolone, Prednisolone F