Learn More
Selleck Chemical LLC Dexamethasone S1322-1g
SDP
Catalog No. 503108270
Encompass_Preferred
Dexamethasone is a potent synthetic member of the glucocorticoid class of steroid drugs and an interleukin receptor modulator that has anti-inflammatory and immunosuppressant effects Dexamethasone induces autophagy and mitophagy Dexamethasone is tested in hospitalized patients with COVID-19 and is found to have benefits for critically ill patients
Catalog No. 50-310-8270 Supplier Selleck Chemical LLC Supplier No. S13221G
Dexamethasone is a potent synthetic member of the glucocorticoid class of steroid drugs and an interleukin receptor modulator that has anti-inflammatory and immunosuppressant effects Dexamethasone induces autophagy and mitophagy Dexamethasone is tested in hospitalized patients with COVID-19 and is found to have benefits for critically ill patients