Learn More
Enzo Life Sciences Dexamethasone, (1g), CAS Number: 50-02-2
SDP
Supplier:  Enzo Life Sciences BMLEI1260001
Encompass_Preferred
Glucocorticoid receptor ligand. Alternative Name: 9alpha-Fluoro-16alpha-methylprednisolone. Formula: C22H29FO5. MW: 392.5. CAS Number: 50-02-2. MI: 14: 2943. Purity: ≥98% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO (25mg/ml), 100% ethanol (25mg/1.6ml) or methanol; insoluble in water. Use/Stability: Stable for up to 5 years after receipt when stored at +20°C. Stock solutions are stable for up to 3 months at -20°C. Long Term Storage: Ambient
Catalog No. NC9139119