Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dexamethasone-21-acetate, 99.7%, MP Biomedicals™

Catalog No. ICN19004194 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19004194 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19004194 Supplier MP Biomedicals, Inc Supplier No. 0219004194

missing translation for 'validMainframeMsg'

Specifications

CAS 1177-87-3
Color White
Assay Percent Range 99.7%
Molecular Formula C24H31FO6
Quantity 5 g
MDL Number MFCD00027407
Synonym dexamethasone acetate, 11b,16a-9-fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione 21-acetate
InChI Key AKUJBENLRBOFTD-RPRRAYFGSA-N
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COC(C)=O
IUPAC Name 2-[(1R,2R,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-2,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
Molecular Weight (g/mol) 434.50
PubChem CID 40469526
Formula Weight 434.5
Physical Form Powder
Chemical Name or Material Dexamethasone-21-acetate
Show More Show Less
Recommended Storage Store at + 4°C
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.