Learn More
Medchemexpress LLC Dexamethasone-4,6α,21,21-d4 | 2483831-63-4 | MFCD01861649 | 99.0% | 396.49 | 1 MG
SDP
Catalog No. 5000360959
Encompass_Preferred
Dexamethasone-4,6α,21,21-d4 is the deuterium labeled Dexamethasone-4,6α,21,21. This compound acts as a glucocorticoid receptor agonist. It significantly decreases CD11b, CD18, and CD62L expression on neutrophils, and CD11b and CD18 expression on monocytes. It is also effective in the control of COVID-19 infection and inhibits the production of exosomes containing inflammatory microRNA-155 in lipopolysaccharide-induced macrophage inflammatory responses.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-60959 Supplier Medchemexpress LLC Supplier No. HY14648S31MG
Dexamethasone-4,6α,21,21-d4 is the deuterium labeled Dexamethasone-4,6α,21,21. This compound acts as a glucocorticoid receptor agonist. It significantly decreases CD11b, CD18, and CD62L expression on neutrophils, and CD11b and CD18 expression on monocytes. It is also effective in the control of COVID-19 infection and inhibits the production of exosomes containing inflammatory microRNA-155 in lipopolysaccharide-induced macrophage inflammatory responses.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.