Learn More
BIOGEMS INTERNATIONAL INC Dexamethasone 500mg
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 5000222500MG
Encompass
Dexamethasone is a synthetic anti-inflammatory and immunosuppressant steroid. It has a stronger affinity for the glucocorticoid receptor than the natural hydrocortisone ligand. Dexamethasone is reported to inhibit mature dendritic cell differentiation and to promote the differentiation of human mesenchymal cells.
Catalog No. 50-153-0165