Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dexamethasone, 96%, Thermo Scientific Chemicals

Catalog No. AC230302500
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC230302500 250 mg Glass bottle
AC230300010 1 g Glass bottle
AC230300050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC230302500 Supplier Thermo Scientific Chemicals Supplier No. 230302500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Dexamethasone, CAS# 50-02-2, is a synthetic adrenal corticosteroid that shows potent anti-inflammatory activity.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• Dexamethasone is a synthetic adrenal glucocorticoid that modulates carbohydrate metabolism, inhibits adrenocorticotropic hormone secretion, and has pronounced anti-inflammatory activity
• Dexamethasone can bind to specific nuclear steroid receptors, inhibiting pro-inflammatory signals, promoting anti-inflammatory signals, and interfering with NF-kB activation and apoptotic pathways

Applications

• It can be used to study apoptosis, cell signaling pathways, and gene expression
• This compound also shows adrenergic, antiemetic, and antineoplastic activities
• It has been used as an important regulator of cellular proliferation and differentiation in vitro human cell cultures
• Dexamethasone has been applied in culture media to promote the differentiation of adipose-derived stem cells into osteogenic tissue
• It is used in in vitro stimulation of primary normal human bronchial epithelial cells (NHBE) and airway smooth muscle (ASM)
• This compound is used for its ability to regulate T cell survival, growth, and differentiation.
• It can also inhibit the induction of nitric oxide synthase.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-02-2
Molecular Formula C22H29FO5
Molecular Weight (g/mol) 392.47
MDL Number MFCD00064136
InChI Key UREBDLICKHMUKA-CXSFZGCWSA-N
Synonym dexamethasone, decadron, dexamethazone, maxidex, decaspray, desametasone, hexadecadrol, dexacort, dexasone, hexadrol
PubChem CID 5743
ChEBI CHEBI:41879
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO

Specifications

Melting Point 255.0°C to 261.0°C
Color White
Loss on Drying 0.5% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 250 mg
Merck Index 15, 2945
Specific Rotation + 75.00
Solubility Information Solubility in water: 10mg/100mL (25 c). Other solubilities: soluble in acetone,ethanol and chloroform
Spectrophotometry 95% min. (on dried substance)
IUPAC Name (8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
Formula Weight 392.46
Percent Purity 96%
Related Substances (HPLC) Passes test
Physical Form Crystalline Powder
Chemical Name or Material Dexamethasone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Suspected of causing cancer.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-003-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.