Learn More
Medchemexpress LLC Dexamethasone acetate | 1177-87-3 | 98.2% | 434.50 | 1 G
SDP
Catalog No. 5000360244
Encompass_Preferred
Dexamethasone acetate is a glucocorticoid receptor agonist that significantly reduces expression of CD11b, CD18, and CD62L on neutrophils, and CD11b and CD18 on monocytes. It is effective in controlling COVID-19 infection and inhibits inflammatory microRNA-155 exosome production in lipopolysaccharide-induced macrophage responses.
  • Functions as a glucocorticoid receptor agonist.
  • Decreases CD11b, CD18, and CD62L expression on neutrophils and monocytes.
  • Effective in controlling COVID-19 infection.
  • Inhibits inflammatory microRNA-155 exosome production in macrophage responses.
  • Targets: Glucocorticoid receptor, autophagy, mitophagy, SARS-CoV, and the complement system.
  • Applicable in inflammation/immunology, endocrinology, and cancer research.
Catalog No. 50-003-60244 Supplier Medchemexpress LLC Supplier No. HY14648A1G
Dexamethasone acetate is a glucocorticoid receptor agonist that significantly reduces expression of CD11b, CD18, and CD62L on neutrophils, and CD11b and CD18 on monocytes. It is effective in controlling COVID-19 infection and inhibits inflammatory microRNA-155 exosome production in lipopolysaccharide-induced macrophage responses.
  • Functions as a glucocorticoid receptor agonist.
  • Decreases CD11b, CD18, and CD62L expression on neutrophils and monocytes.
  • Effective in controlling COVID-19 infection.
  • Inhibits inflammatory microRNA-155 exosome production in macrophage responses.
  • Targets: Glucocorticoid receptor, autophagy, mitophagy, SARS-CoV, and the complement system.
  • Applicable in inflammation/immunology, endocrinology, and cancer research.