Learn More
Apexbio Technology LLC Dexamethasone acetate 1177-87-3 5g
SDP
Catalog No. 5000063656
Encompass_Preferred
Dexamethasone acetate a synthetic glucocorticoid acts by binding to glucocorticoid receptors influencing gene transcription to suppress inflammatory and immune responses By modulating cytokine production and inhibiting interleukin (IL) signaling pathways dexamethasone acetate serves as an interleukin receptor modulator Typically applied in research related to inflammation autoimmune diseases and cytokine-mediated cellular events it is extensively utilized in functional assays to understand receptor-ligand interactions signal transduction and therapeutic mechanisms Studies report variable IC50 values depending on cell type and assay conditions with typical IC50 ranges documented from 1-10 nM in receptor binding assays providing a reference concentration range for experimental designs
Catalog No. 50-000-63656 Supplier Apexbio Technology LLC Supplier No. B19265000
Dexamethasone acetate a synthetic glucocorticoid acts by binding to glucocorticoid receptors influencing gene transcription to suppress inflammatory and immune responses By modulating cytokine production and inhibiting interleukin (IL) signaling pathways dexamethasone acetate serves as an interleukin receptor modulator Typically applied in research related to inflammation autoimmune diseases and cytokine-mediated cellular events it is extensively utilized in functional assays to understand receptor-ligand interactions signal transduction and therapeutic mechanisms Studies report variable IC50 values depending on cell type and assay conditions with typical IC50 ranges documented from 1-10 nM in receptor binding assays providing a reference concentration range for experimental designs