Learn More
Cayman Chemical Dexamethasone, 50-02-2, 500 mg
SDP
Supplier:  Cayman Chemical 11015500
Encompass_Preferred
Molecular Formula C22H29FO5, Purity ≥98%, Formula Weight 392.5, Synonyms: 9-fluoro-11β,17,21-trihydroxy-16α-methyl-pregna-1,4-diene-3,20-dione, MK-125, NSC 34521; A synthetic glucocorticoid that binds the human glucocorticoid receptor with a higher affinity than a natural ligand, cortisol (Kd = 5 vs. 17 nM, respectively); has anti-inflammatory effects.
Catalog No. NC0529870