Learn More
Medchemexpress LLC Dexamethasone-d4 | 2305607-27-4 | 98.9% | 396.49 | 1 MG
SDP
Catalog No. 5000363652
Encompass_Preferred
Dexamethasone-d4 is a deuterium-labeled Dexamethasone, which acts as a glucocorticoid receptor agonist. It significantly decreases CD11b, CD18, and CD62L expression on neutrophils, and CD11b and CD18 expression on monocytes. Dexamethasone is highly effective in controlling COVID-19 infection and inhibits the production of exosomes containing inflammatory microRNA-155 in lipopolysaccharide-induced macrophage inflammatory responses. This compound is versatile for various analytical applications.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-63652 Supplier Medchemexpress LLC Supplier No. HY14648S21MG
Dexamethasone-d4 is a deuterium-labeled Dexamethasone, which acts as a glucocorticoid receptor agonist. It significantly decreases CD11b, CD18, and CD62L expression on neutrophils, and CD11b and CD18 expression on monocytes. Dexamethasone is highly effective in controlling COVID-19 infection and inhibits the production of exosomes containing inflammatory microRNA-155 in lipopolysaccharide-induced macrophage inflammatory responses. This compound is versatile for various analytical applications.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS