Learn More
Selleck Chemical LLC Dexamethasone 50mg 50-02-2
SDP
Supplier:  Selleck Chemical LLC S132250MG
Encompass_Preferred
Dexamethasone is a potent synthetic member of the glucocorticoid class of steroid drugs, and an interleukin receptor modulator that has anti-inflammatory and immunosuppressant effects. Dexamethasone induces autophagy and mitophagy. Dexamethasone is tested in hospitalized patients with COVID-19 and is found to have benefits for critically ill patients. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0854626