Learn More
Apexbio Technology LLC Dexamethasone (DHAP) 50-02-2 5g
SDP
Supplier:  Apexbio Technology LLC A23245000
Encompass_Preferred
Dexamethasone (DHAP) is a synthetic glucocorticoid commonly utilized in biomedical research due to its potent anti-inflammatory function and immunomodulatory effects It exerts its mechanism through glucocorticoid receptor-mediated transcriptional modulation resulting in reduced transcriptional activity of NF- B signaling pathways in dendritic cells (DC) thus inhibiting DC maturation and activation This compound is frequently employed in experiments investigating immune regulation inflammation and cell differentiation processes including induction of differentiation in human mesenchymal stem cells (MSCs) Additionally dexamethasone is used in cancer cell biology research as it promotes autophagy in acute lymphoblastic leukemia-derived cell lines Reported IC50 values for dexamethasone-mediated inhibition of cellular inflammation responses typically range from 2 50 nM depending on specific cellular models and experimental conditions
Catalog No. 50-000-62995