Learn More
Medchemexpress LLC Dexamethasone (GMP) | 50-02-2 | MFCD00064136 | >99.8% | 392.46 | 50 MG
SDP
Catalog No. 5000363651
Encompass_Preferred
Dexamethasone (Hexadecadrol) (GMP) is a glucocorticoid receptor agonist produced using GMP guidelines. It functions as an auxiliary reagent for cell therapy manufacture.
  • Increases myotubes formation and cytoplasmic lipid droplets accumulation
  • Induces osteogenesis of chick periosteum cells
  • Induces osteoblastic differentiation
  • Stimulates differentiation of odontoblast-like cells
Catalog No. 50-003-63651 Supplier Medchemexpress LLC Supplier No. HY14648G50MG
Dexamethasone (Hexadecadrol) (GMP) is a glucocorticoid receptor agonist produced using GMP guidelines. It functions as an auxiliary reagent for cell therapy manufacture.
  • Increases myotubes formation and cytoplasmic lipid droplets accumulation
  • Induces osteogenesis of chick periosteum cells
  • Induces osteoblastic differentiation
  • Stimulates differentiation of odontoblast-like cells