Learn More
Medchemexpress LLC Dexamethasone impuri 5mg | 37927-01-8 | 5 MG
SDP
Catalog No. 5000229988
Encompass_Preferred
Dexamethasone acid (CAS 37927-01-8) is an identified impurity of dexamethasone used as a reference standard for analytical and stability testing. It is a solid with molecular formula C21H27FO5 and molecular weight ~378.43 g/mol, typically supplied in small packs for laboratory analysis.
  • use as a reference standard for impurity profiling and method validation.
  • suitable for HPLC analysis and chromatographic comparison.
  • typical reported purity greater than 95% by HPLC.
  • supplied in small quantities for analytical and quality-control use.
Catalog No. 50-002-29988 Supplier Medchemexpress LLC Supplier No. HY1144325MG
Dexamethasone acid (CAS 37927-01-8) is an identified impurity of dexamethasone used as a reference standard for analytical and stability testing. It is a solid with molecular formula C21H27FO5 and molecular weight ~378.43 g/mol, typically supplied in small packs for laboratory analysis.
  • use as a reference standard for impurity profiling and method validation.
  • suitable for HPLC analysis and chromatographic comparison.
  • typical reported purity greater than 95% by HPLC.
  • supplied in small quantities for analytical and quality-control use.