Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dexamethasone Impurity Standard, MilliporeSigma™ Supelco™

Catalog No. 111002047 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2047 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2047 Supplier MilliporeSigma Supelco Supplier No. BP811
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
TRUSTED_SUSTAINABILITY

Specifications

CAS 50-02-2
Linear Formula C22H29 FO5
MDL Number MFCD00064136
Melting Point 262°C to 264°C
Synonym Dexamethasone; (11 beta,16 alpha)-9-Fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione; 9 alpha-Fluoro-16 alpha-methyl-11 beta,17 alpha,21-trihydroxy-1,4-pregnadiene-3,20-dione; 9 alpha-Fluoro-16 alpha-methylprednisolone; Prednisolone F
Type Pharmacopeia and Metrological Institutes Standards
Quantity 25 mg
Formula Weight 392.46
Grade Pharmaceutical Primary Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.