Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dexamethasone, Micronized, USP, 97-102%, Spectrum™ Chemical
SDP

Catalog No. 18602055 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-055 1 g Amber Glass Bottle
18-602-056 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-602-055 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. DE1211GM

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

DE121, 50-02-2, C22H29FO5

Spectrum™ Chemical Dexamethasone, Micronized, USP is corticosteroid that has strong anti-inflammatory and immunosuppressant effects. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 50-02-2
Molecular Formula C22H29FO5
Molecular Weight (g/mol) 392.47
MDL Number MFCD00064136
InChI Key UREBDLICKHMUKA-CXSFZGCWSA-N
IUPAC Name (1R,2R,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-1-(2-hydroxyacetyl)-2,9a,11a-trimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Loss on Drying 0.5%
Melting Range 260°C to 264°C
Packaging Amber Glass Bottle
Quantity 1 g
Specific Rotation +72° to +80°
Percent Purity 97 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Dexamethasone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.