Learn More
Medchemexpress LLC HY-128922 100mg , Dexamethasone palmitate CAS:14899-36-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY128922100MG
Encompass_Preferred
HY-128922 100mg Dexamethasone palmitate CAS: 14899-36-6 Dexamethasone palmitate (DXP) is a prodrug of Dexamethasone (HY-14648). Dexamethasone palmitate can be used for the research of inflammation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0023