Learn More
Medchemexpress LLC Dexamethasone palmitate | 14899-36-6 | 99.68% | 1 ML
SDP
Catalog No. 5000340485
Encompass_Preferred
Dexamethasone palmitate (DXP) is a prodrug of Dexamethasone (HY-14648) and can be utilized for inflammation research. It has demonstrated in vitro anti-inflammatory effects by decreasing LPS-induced MCP-1 and TNF-α concentration in RAW 264.7 cells. In vivo studies showed that a single intravitreal (IVT) injection resulted in distribution mostly in the vitreous, persisting for up to 9 months, and controlled VEGF-induced vascular hyperpermeability.
  • Utilized for inflammation research
  • Decreases LPS-induced MCP-1 and TNF-α concentration
  • Shows sustained distribution in vitreous for up to 9 months
  • Controls VEGF-induced vascular hyperpermeability
Catalog No. 50-003-40485 Supplier Medchemexpress LLC Supplier No. HY12892210MM/1ML
Dexamethasone palmitate (DXP) is a prodrug of Dexamethasone (HY-14648) and can be utilized for inflammation research. It has demonstrated in vitro anti-inflammatory effects by decreasing LPS-induced MCP-1 and TNF-α concentration in RAW 264.7 cells. In vivo studies showed that a single intravitreal (IVT) injection resulted in distribution mostly in the vitreous, persisting for up to 9 months, and controlled VEGF-induced vascular hyperpermeability.
  • Utilized for inflammation research
  • Decreases LPS-induced MCP-1 and TNF-α concentration
  • Shows sustained distribution in vitreous for up to 9 months
  • Controls VEGF-induced vascular hyperpermeability