Learn More
Medchemexpress LLC Dexamethasone palmitate | 14899-36-6 | 99.7% | 10 MG
SDP
Catalog No. 5000338576
Encompass_Preferred
Dexamethasone palmitate (DXP) is a proagent of Dexamethasone and can be used for inflammation research. This compound has demonstrated significant biological activity both in vitro and in vivo.
  • Affects cytokine concentration of RAW 264.7 cells in vitro.
  • Shows in vitro anti-inflammatory effects by decreasing LPS-induced MCP-1 and TNF-α concentration in RAW 264.7 cells.
  • Exhibits different distribution in ocular tissue at various times during in vivo studies.
  • Affects VEGF-induced vascular hyperpermeability in vivo.
Catalog No. 50-003-38576 Supplier Medchemexpress LLC Supplier No. HY12892210MG
Dexamethasone palmitate (DXP) is a proagent of Dexamethasone and can be used for inflammation research. This compound has demonstrated significant biological activity both in vitro and in vivo.
  • Affects cytokine concentration of RAW 264.7 cells in vitro.
  • Shows in vitro anti-inflammatory effects by decreasing LPS-induced MCP-1 and TNF-α concentration in RAW 264.7 cells.
  • Exhibits different distribution in ocular tissue at various times during in vivo studies.
  • Affects VEGF-induced vascular hyperpermeability in vivo.