Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dexamethasone palmitate | 14899-36-6 | 99.7% | 50 MG
SDP

Catalog No. 5000340643
Encompass_Preferred

Dexamethasone palmitate (DXP) is a prodrug of Dexamethasone, primarily utilized in inflammation research. It has demonstrated in vitro anti-inflammatory effects by decreasing LPS-induced MCP-1 and TNF-α concentrations in RAW 264.7 cells. In vivo studies in New Zealand White rabbits have shown that Dexamethasone palmitate, administered via IVT injection, distributes significantly in the vitreous humor and can control VEGF-induced vascular hyperpermeability.

  • Appearance: Solid, white to off-white
  • Purity: 99.68%
  • Molecular weight: 630.87
  • Formula: C38H59FO6
  • Primarily for inflammation research
  • Demonstrates in vitro anti-inflammatory effects (decreases LPS-induced MCP-1 and TNF-α)
  • In vivo studies show significant distribution in vitreous humor and control of VEGF-induced vascular hyperpermeability

Catalog No. 50-003-40643 Supplier Medchemexpress LLC Supplier No. HY12892250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dexamethasone palmitate (DXP) is a prodrug of Dexamethasone, primarily utilized in inflammation research. It has demonstrated in vitro anti-inflammatory effects by decreasing LPS-induced MCP-1 and TNF-α concentrations in RAW 264.7 cells. In vivo studies in New Zealand White rabbits have shown that Dexamethasone palmitate, administered via IVT injection, distributes significantly in the vitreous humor and can control VEGF-induced vascular hyperpermeability.

  • Appearance: Solid, white to off-white
  • Purity: 99.68%
  • Molecular weight: 630.87
  • Formula: C38H59FO6
  • Primarily for inflammation research
  • Demonstrates in vitro anti-inflammatory effects (decreases LPS-induced MCP-1 and TNF-α)
  • In vivo studies show significant distribution in vitreous humor and control of VEGF-induced vascular hyperpermeability

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.