Learn More
Selleck Chemical LLC Dexamethasone palmitate S5184-5mg
SDP
Catalog No. 503111886
Encompass_Preferred
Dexamethasone palmitate (DXP) a lipophilic prodrug of Dexamethasone (DXM) is a glucocorticoid receptor agonist with a 47-fold lower affinity for the glucocorticoid receptor than DXM Dexamethasone palmitate (DXP) exhibits anti-inflammatory activity
Catalog No. 50-311-1886 Supplier Selleck Chemical LLC Supplier No. S51845MG
Dexamethasone palmitate (DXP) a lipophilic prodrug of Dexamethasone (DXM) is a glucocorticoid receptor agonist with a 47-fold lower affinity for the glucocorticoid receptor than DXM Dexamethasone palmitate (DXP) exhibits anti-inflammatory activity