Learn More
Medchemexpress LLC HY-128922 5mg Medchemexpress, Dexamethasone palmitate CAS:14899-36-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1289225MG
Encompass_Preferred
Medchemexpress, HY-128922 5mg Dexamethasone palmitate CAS:14899-36-6 Dexamethasone palmitate (DXP) is a prodrug of Dexamethasone, which is a glucocorticoid receptor agonist. Dexamethasone palmitate (DXP) has a 47-fold lower affinity for the glucocorticoid receptor than Dexamethasone. Anti-inflammatory agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9784