Learn More
Medchemexpress LLC Dexamethasone palmitate-d31 | 14899-36-6 | 99.3% | 2.5 MG
SDP
Catalog No. 5000340442
Encompass_Preferred
Dexamethasone palmitate-d31 is an isotope-labeled compound, specifically a deuterium-labeled version of Dexamethasone palmitate, which is a proagent of Dexamethasone. This compound is primarily utilized in research contexts, particularly for studies related to inflammation.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis.
  • Compatible with analytical techniques such as NMR, GC-MS, and LC-MS.
Catalog No. 50-003-40442 Supplier Medchemexpress LLC Supplier No. HY128922S2.5MG
Dexamethasone palmitate-d31 is an isotope-labeled compound, specifically a deuterium-labeled version of Dexamethasone palmitate, which is a proagent of Dexamethasone. This compound is primarily utilized in research contexts, particularly for studies related to inflammation.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis.
  • Compatible with analytical techniques such as NMR, GC-MS, and LC-MS.