Learn More
AdipoGen Dexamethasone phosphate . disodium salt (500 mg)
SDP
Supplier:  AdipoGen AGCR13744M500
Encompass_Preferred
Chemical. CAS: 2392-39-4. Formula: C22H28FO8P . 2Na. MW: 470.4 . 46.0. Dexamethasone phosphate is a water-soluble form of the synthetic glucocorticoid dexamethasone (AG-CR1-3742). Dexamethasone is a potent anti-inflammatory and immunosuppressive agent useful in a variety of inflammatory and autoimmune conditions. This synthetic glucocorticoid acts as an agonist of human glucocorticoid receptor (GR) and binds with a higher affinity than the natural ligand cortisol (Kd = 5nM versus 17nM, respectively). Through GR activation, dexamethasone has both transactivating and transrepressing effects on gene expression, resulting in anti-inflammatory properties. The anti-inflammatory effects are complex, but primarily via inhibition of inflammatory cells and suppression of expression of inflammatory mediators. Dexamethasone has been shown to regulate T cell survival, growth and differentiation and improves survival rates of hospitalized patients with COVID-19 by inhibiting cytokine storm.
Catalog No. 50-205-4186