Learn More
Apexbio Technology LLC Dexamethasone phosphate disodium 2392-39-4 100mg
SDP
Catalog No. 5000089843
Encompass_Preferred
Dexamethasone phosphate disodium (CAS 2392-39-4) is a small-molecule agonist targeting the glucocorticoid receptor It is designed to modulate glucocorticoid-mediated signaling pathways thereby regulating immune and inflammatory responses Dexamethasone phosphate disodium exerts its biological activity primarily by binding to the glucocorticoid receptor initiating nuclear translocation and influencing gene transcription to suppress the expression and secretion of pro-inflammatory cytokines and chemokines Based on these pharmacological properties dexamethasone phosphate disodium holds research potential in studying inflammatory conditions immune response processes and associated signaling cascades in vitro and in vivo
Catalog No. 50-000-89843 Supplier Apexbio Technology LLC Supplier No. C6550100
Dexamethasone phosphate disodium (CAS 2392-39-4) is a small-molecule agonist targeting the glucocorticoid receptor It is designed to modulate glucocorticoid-mediated signaling pathways thereby regulating immune and inflammatory responses Dexamethasone phosphate disodium exerts its biological activity primarily by binding to the glucocorticoid receptor initiating nuclear translocation and influencing gene transcription to suppress the expression and secretion of pro-inflammatory cytokines and chemokines Based on these pharmacological properties dexamethasone phosphate disodium holds research potential in studying inflammatory conditions immune response processes and associated signaling cascades in vitro and in vivo