Learn More
Medchemexpress LLC Dexamethasone phosphate disodium | 2392-39-4 | 99.9% | 1 G
SDP
Catalog No. 5000412224
Encompass_Preferred
Dexamethasone phosphate disodium is an orally active Glucocorticoid receptor agonist.
  • Orally active Glucocorticoid receptor agonist.
  • Soluble in water (≥ 100 mg/mL).
  • Soluble in DMSO (1 mg/mL).
  • Cited in 15 publications, with 8 specifically using this product.
Catalog No. 50-004-12224 Supplier Medchemexpress LLC Supplier No. HYB1829A1G
Dexamethasone phosphate disodium is an orally active Glucocorticoid receptor agonist.
  • Orally active Glucocorticoid receptor agonist.
  • Soluble in water (≥ 100 mg/mL).
  • Soluble in DMSO (1 mg/mL).
  • Cited in 15 publications, with 8 specifically using this product.