Learn More
Medchemexpress LLC Dexamethasone phosphate disodium | 2392-39-4 | 99.9% | 1 ML
SDP
Catalog No. 5000409776
Encompass_Preferred
Dexamethasone phosphate disodium is an orally active Glucocorticoid receptor agonist. It regulates several transcription factors, including activator protein-1, nuclear factor-AT, and nuclear factor-kB, leading to the activation and repression of key genes involved in the inflammatory response.
  • Potently inhibits granulocyte-macrophage colony stimulating factor (GM-CSF) release from A549 cells with EC50 of 2.2 nM.
  • Induces transcription of the β2-receptor (EC50=36 nM).
  • Inhibits 3xκB (NF-κB, IκBα, and I-κBβ) with IC50=0.5 nM.
  • Effectively inhibits lipopolysaccharide (LPS)-induced inflammation.
  • Decreases recruitment of granulocytes and spontaneous production of oxygen radicals.
Catalog No. 50-004-09776 Supplier Medchemexpress LLC Supplier No. HYB1829A10MM/1ML
Dexamethasone phosphate disodium is an orally active Glucocorticoid receptor agonist. It regulates several transcription factors, including activator protein-1, nuclear factor-AT, and nuclear factor-kB, leading to the activation and repression of key genes involved in the inflammatory response.
  • Potently inhibits granulocyte-macrophage colony stimulating factor (GM-CSF) release from A549 cells with EC50 of 2.2 nM.
  • Induces transcription of the β2-receptor (EC50=36 nM).
  • Inhibits 3xκB (NF-κB, IκBα, and I-κBβ) with IC50=0.5 nM.
  • Effectively inhibits lipopolysaccharide (LPS)-induced inflammation.
  • Decreases recruitment of granulocytes and spontaneous production of oxygen radicals.