Learn More
Medchemexpress LLC Dexamethasone phosphate disodium | 2392-39-4 | 99.9% | 5 G
SDP
Catalog No. 5000410796
Encompass_Preferred
Dexamethasone phosphate disodium is an orally active Glucocorticoid receptor agonist.
  • Regulates several transcription factors, including activator protein-1, nuclear factor-AT, and nuclear factor-kB, leading to the activation and repression of key genes involved in the inflammatory response.
  • Potently inhibits granulocyte-macrophage colony stimulating factor (GM-CSF) release from A549 cells with EC50 of 2.2 nM.
  • Induces transcription of the β2-receptor at 10-100 fold higher concentrations than the inhibition of GM-CSF release (EC50=36 nM).
  • Inhibits 3xκB (NF-κB, IκBα, and I-κBβ) with IC50=0.5 nM, which is associated with inhibition of GM-CSF release.
  • Efficiently inhibits lipopolysaccharide (LPS)-induced inflammation at a dose of 2x5 mg/kg.
Catalog No. 50-004-10796 Supplier Medchemexpress LLC Supplier No. HYB1829A5G
Add to Cart
Add to Cart
Dexamethasone phosphate disodium is an orally active Glucocorticoid receptor agonist.
  • Regulates several transcription factors, including activator protein-1, nuclear factor-AT, and nuclear factor-kB, leading to the activation and repression of key genes involved in the inflammatory response.
  • Potently inhibits granulocyte-macrophage colony stimulating factor (GM-CSF) release from A549 cells with EC50 of 2.2 nM.
  • Induces transcription of the β2-receptor at 10-100 fold higher concentrations than the inhibition of GM-CSF release (EC50=36 nM).
  • Inhibits 3xκB (NF-κB, IκBα, and I-κBβ) with IC50=0.5 nM, which is associated with inhibition of GM-CSF release.
  • Efficiently inhibits lipopolysaccharide (LPS)-induced inflammation at a dose of 2x5 mg/kg.