Learn More
Medchemexpress LLC Dexamethasone phosphate disodium | 2392-39-4 | 99.94% | 516.40 | 500 MG
SDP
Catalog No. 5000365906
Encompass_Preferred
Dexamethasone phosphate disodium is an orally active Glucocorticoid receptor agonist. It regulates various transcription factors, including activator protein-1, nuclear factor-AT, and nuclear factor-kB, to influence inflammatory response genes.
  • Potently inhibits granulocyte-macrophage colony stimulating factor (GM-CSF) release from A549 cells
  • Induces transcription of the β2-receptor
  • Inhibits 3xκB (NF-κB, IκBα, and I-κBβ) activity
  • Efficiently inhibits lipopolysaccharide (LPS)-induced inflammation in vivo
  • Decreases recruitment of granulocytes and spontaneous production of oxygen radicals in vivo
Catalog No. 50-003-65906 Supplier Medchemexpress LLC Supplier No. 500MGHYB1829A
Dexamethasone phosphate disodium is an orally active Glucocorticoid receptor agonist. It regulates various transcription factors, including activator protein-1, nuclear factor-AT, and nuclear factor-kB, to influence inflammatory response genes.
  • Potently inhibits granulocyte-macrophage colony stimulating factor (GM-CSF) release from A549 cells
  • Induces transcription of the β2-receptor
  • Inhibits 3xκB (NF-κB, IκBα, and I-κBβ) activity
  • Efficiently inhibits lipopolysaccharide (LPS)-induced inflammation in vivo
  • Decreases recruitment of granulocytes and spontaneous production of oxygen radicals in vivo