Learn More
Medchemexpress LLC Dexamethasone phosphate disodium (Standard) | 2392-39-4 | 99.88% | 100 MG
SDP
Catalog No. 5000412225
Encompass_Preferred
Dexamethasone phosphate (disodium) (Standard) is the analytical standard of Dexamethasone phosphate (disodium). This product is intended for research and analytical applications. Dexamethasone phosphate (Dexamethasone 21-phosphate) disodium is an orally active Glucocorticoid receptor agonist.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-004-12225 Supplier Medchemexpress LLC Supplier No. HYB1829AR100MG
Dexamethasone phosphate (disodium) (Standard) is the analytical standard of Dexamethasone phosphate (disodium). This product is intended for research and analytical applications. Dexamethasone phosphate (Dexamethasone 21-phosphate) disodium is an orally active Glucocorticoid receptor agonist.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.