Learn More
Medchemexpress LLC Dexamethasone phosphate disodium (Standard) | 2392-39-4 | 99.88% | 25 MG
SDP
Catalog No. 5000413850
Encompass_Preferred
Dexamethasone phosphate disodium (Standard) is the analytical standard of Dexamethasone phosphate (disodium). It is an orally active Glucocorticoid receptor agonist intended for research and analytical applications. It is commonly used as a reference standard in qualitative, quantitative, and methodological research experiments.
  • Used in HPLC, GC, and MS applications
  • Orally active glucocorticoid receptor agonist
  • Reference standard for research and analytical use
Catalog No. 50-004-13850 Supplier Medchemexpress LLC Supplier No. 25MGHYB1829AR
Dexamethasone phosphate disodium (Standard) is the analytical standard of Dexamethasone phosphate (disodium). It is an orally active Glucocorticoid receptor agonist intended for research and analytical applications. It is commonly used as a reference standard in qualitative, quantitative, and methodological research experiments.
  • Used in HPLC, GC, and MS applications
  • Orally active glucocorticoid receptor agonist
  • Reference standard for research and analytical use