Learn More
Cayman Chemical Dexamethasone Phosphate sodi
SDP
Catalog No. 502984771
Encompass_Preferred
A glucocorticoid; reduces LPS-induced production of RANTES, TGF-β1, and NO in BV-2 microglia from 1-4 µM; reverses LPS-induced decreases in IL-10 levels in, and inhibits LPS-induced migration of, BV-2 microglia at 4 µM; decreases motor activity and reduces adrenal weight and plasma corticosterone levels in rats when administered in the drinking water at 5 UG/ml; reduces joint swelling in a rat model of collagen-induced arthritis at 1.6 mg/kg per day for a minimum of seven days; reduces pruritis and the clinical lesion score in cats with hypersensitivity dermatitis
Catalog No. 50-298-4771 Supplier Cayman Chemical Supplier No. 20340100 MG
A glucocorticoid; reduces LPS-induced production of RANTES, TGF-β1, and NO in BV-2 microglia from 1-4 µM; reverses LPS-induced decreases in IL-10 levels in, and inhibits LPS-induced migration of, BV-2 microglia at 4 µM; decreases motor activity and reduces adrenal weight and plasma corticosterone levels in rats when administered in the drinking water at 5 UG/ml; reduces joint swelling in a rat model of collagen-induced arthritis at 1.6 mg/kg per day for a minimum of seven days; reduces pruritis and the clinical lesion score in cats with hypersensitivity dermatitis