Learn More
STEMCELL Technologies Dexamethasone, Size: 500 mg
SDP
Supplier:  STEMCELL Technologies 72092
Encompass
Dexamethasone is a synthetic glucocorticoid, similar to the natural glucocorticoid hydrocortisone. Dexamethasone has an increased affinity for glucocorticoid receptors when compared to the natural hydrocortisone ligand (Kd = 5 nM vs 17 nM).REPROGRAMMING· Promotes transdifferentiation of hepatocytes from mouse pancreatic cells (Shen et al.).DIFFERENTIATION· Promotes osteogenic, adipogenic, and chondrogenic differentiation of human mesenchymal cells (Jaiswal et al., Mackay et al., Pittenger et al.).· Promotes osteogenic, adipogenic, and chondrogenic differentiation of mouse mesenchymal cells (Tropel et al.).· Promotes differentiation of mature hepatocytes from mouse and human embryonic stem (ES) cells (Cai et al., Kubo et al.).· Promotes maturation of fetal mouse hepatocytes (Kamiya et al.).
Catalog No. 50-197-5882